Learn More
Invitrogen™ ARF1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA1127
Product Code. 15344791
  • 556.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
Description
Description
PA1-127 has been successfully used in Western blot in human, mouse, and canine samples, and in immunofluorescence and immunoprecipitation applications in human samples.
ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ARF1
Polyclonal
Unconjugated
ARF1
ADP ribosylation factor 1; ADP ribosylation factor 1 L homeolog; ADP-ribosylation factor 1; Arf1; arf1.L; arf2; wu:fb33e02; wu:fb78h01; XELAEV_18043369mg
Rabbit
Antigen affinity chromatography
RUO
100856517, 11840, 375
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 30% glycerol, 1mg/mL BSA and 0.05% sodium azide
P84077, P84078
ARF1
Synthetic peptide DWLSNQLRNQK.
100 μg
Primary
Human, Mouse, Canine