missing translation for 'onlineSavingsMsg'
Learn More

ARF1 Antibody (ARFS 1A9/5), DyLight 650, Novus Biologicals™

Product Code. 18098773 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18098773 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18098773 Supplier Novus Biologicals Supplier No. NB1002635C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARF1 Monoclonal specifically detects ARF1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARF1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone ARFS 1A9/5
Conjugate DyLight 650
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ADP-ribosylation factor 1
Gene Symbols ARF1
Host Species Mouse
Immunogen C-terminal Human ARF1 peptide SNQLRNQ
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375
Test Specificity ARFS 1A9/5 is Class 1 ARF specific.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.