missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARC Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1G4B5, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | ARC |
|---|---|
| Clone | 1G4B5 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808535
|
Proteintech
66550-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898525
|
Proteintech
66550-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Required for consolidation of synaptic plasticity as well as formation of long-term memory. Regulates endocytosis of AMPA receptors in response to synaptic activity. Required for homeostatic synaptic scaling of AMPA receptors. Localized in: Cytoplasm, cytoskeleton. Endosome. Acrosome. Cell junction, synapse, postsynaptic cell membrane, postsynaptic density. Cell projection, dendrite. Cell projection, dendritic spine. Cell junction, synapse. Associated with the cell cortex of neuronal soma and dendrites. Enriched in postsynaptic density of dendritic spines. Associated with the sperm tail.Specifications
| ARC | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ARC, ARC/ARG3.1, Arg3.1, KIAA0278 | |
| Arc | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1G4B5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Pig, Rat | |
| Q63053, Q7LC44, Q9WV31 | |
| 110260202, 11838, 23237, 54323 | |
| ARC Fusion Protein Ag9445 | |
| Primary | |
| -20°C | |
| ARC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title