Learn More
ARC 239 Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | ARC 239 Dihydrochloride |
| CAS | 55974-42-0 |
| Molecular Formula | C24H29N3O3 • 2(HCl) |
| Quantity | 1 mg |
| Synonym | 2-[2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl]-4,4-dimethyl-1,3-(2H,4H)-isoquinolindione Dihydrochloride, 2-[2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl]-4,4-dimethyl-1,3-(2H,4H)-isoquinolindione Dihydrochloride (9CI), 2-[2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl]-4,4-dimethyl-1,3-(2H,4H)-isoquinolindione Hydrochloride (1:2) |
| InChI Formula | InChI=1S/C24H29N3O3.2ClH/c1-24(2)19-9-5-4-8-18(19)22(28)27(23(24)29)17-14-25-12-15-26(16-13-25)20-10-6-7-11-21(20)30-3;;/h4-11H,12-17H2,1-3H3;2*1H |
| SMILES | COC1=CC=CC=C1N2CCN(CC2)CCN3C(C4=C(C(C)(C3=O)C)C=CC=C4)=O |
| IUPAC Name | 2-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-4,4-dimethylisoquinoline-1,3-dione;dihydrochloride |
| Molecular Weight (g/mol) | 407.5123646 |