missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AquaPro™ Multi-Input Intelligent Process Transmitters

Product Code. p-200087469
Click to view available options
Channel 1:
Aquasensors Datastick
Voltage:
90/240 V
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17521685

Brand: Thermo Scientific™ AP8XXXK

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Streamline process monitoring with the Thermo Scientific™ AquaPro™ Multi-Input Intelligent Transmitter. The AquaPro provides accurate, user-friendly monitoring in-line via a reliable, simplified interface, and can be upgraded in the field to accept up to four inputs.

Streamline your operation with the Thermo Scientific™ AquaPro™ Multi-Input Intelligent Transmitter, with pre-installed sensing card. Featuring the ability to simultaneously display up to four (4) inputs or parameters, the AquaPro provides accurate, user-friendly monitoring of in-line process applications via a compact, simplified interface. From large industrial operations, to small municipal plants, the AquaPro offers the flexibility and versatility required to meet a variety of measurement needs.

Versatile and Flexible
The AquaPro Multi-Input Intelligent Process Analyzer is far more than a 4-wire multi-input meter. It is a sophisticated and highly flexible analysis platform for a wide range of process applications. With up to four inputs or parameters, a userfriendly interface, and easy-to-use menus in seven languages, the AquaPro meets the needs of complex process operations worldwide.

Advantages:

  • Large color display
  • Up to four sensor inputs
  • Up to eight current outputs
  • Up to six alarm/control relays
  • Multiple digital communication protocols
  • Broad sensor compatibility
  • Seven user-selectable languages

AquaPro provides a simple interface to PLC, SCADA, and other PC-based control systems and compatibility with all current industry data communication protocols — MODBUS™ RTU over RS485 or RS232, DeviceNet™, Profibus™, Ethernet/IP, Modbus TCP, and CANopen™. It can be easily updated to support any future protocols using our unique modular design.

AquaPro is fully compatible with a wide range of liquid analytical sensors — both analog and digital — allowing measurement of many parameters, including:

  • pH/ORP
  • Conductivity (including resistivity, concentration, salinity, and TDS)
  • Dissolved oxygen
  • Dissolved ozone
  • Free chlorine
  • Turbidity
  • Suspended solids
TRUSTED_SUSTAINABILITY

Specifications

Analog Outputs Two isolated 0/4-20mA per channel (non-reversable), 0.01mA resolution, linear/log/PID capability
Case Material Polycarbonate
Channels 4 (Customizable with modules)
Communication Details Optional MODBUS RTU or Ethernet/IP
Description Intelligent multi-input transmitter with pre-installed sensing card for digital Datastick sensors
Temperature (Metric) Operating -20°C to +60°C
Type Process Analyzer
Display Type 109.22 mm (4.3 in) high-contrast color graphical display
Enclosure Rating NEMA 4X, IP66
For Use With (Equipment) Aquasensors Datastick digital sensors
Keypad Type Tactile keypad with 7-key menu navigation
Max. Relative Humidity 95%, non-condensing
Mounting Options Wall, panel or pipe
Outputs 2 x 4-20 mA outputs 3 relays 240 VAC 50 Hz single phase
Range Sensor Dependant
Relay Details Three form C relays, 5A @ 250VAC
Resolution Sensor Dependant
Temperature (English) Operating -4°F to +140°F
USB Interface Front-panel accessible USB-A for firmware updates and datalog downloads
Dimensions (L x W x H) 144 x 144 x 128 mm (5.67 x 5.67 x 5.01 in.)
Weight (English) 3 lbs.
Weight (Metric) 1.3 kg
Electrical Requirements 100 to 240 VAC, 9.6W, 50/60 Hz
Voltage 90/240 V
Hazardous Locations Class 1 Div 2, Groups A, B, C, D (in combination with approved sensors)
DIN Size 1/2 DIN
Channel 1 Aquasensors Datastick
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.