missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-97927-0.025ml
This item is not returnable.
View return policy
Description
Aquaporin-3 Polyclonal specifically detects Aquaporin-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Aquaporin-3 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| AQP-3, Aquaglyceroporin-3, aquaporin 3, aquaporin 3 (GIL blood group), aquaporin 3 (Gill blood group), aquaporin-3, GIL, Gill blood group | |
| Rabbit | |
| 32 kDa | |
| 0.025 mL | |
| Membrane Trafficking and Chaperones | |
| 360 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:200 | |
| P47862 | |
| AQP3 | |
| Produced against the C-terminal peptide (Sequence N-CHLEQPPPSTEAENVKLAHMKHKEQI) of rat aquaporin 3 | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects approx 31.5kDa. May detect larger glycosylated bands approx 35-50kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction