missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aquaporin 2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13218038
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13218038 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13218038 Supplier Invitrogen™ Supplier No. PA522865

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In Western blot, bands can be seen ~28 kDa for the non-glycosylated form and ~37 kDa for the glycosylated form. Suggested positive control: rat kidney, renal inner medulla.

Aquaporin 2 (AQP2) is a hormonally regulated water channel located in the renal collecting duct. Mutations in the AQP2 gene cause hereditary nephrogenic diabetes insipidus in humans. A vasopressin induced cAMP increase results in the phosphorylation of AQP2 at serine-256 and its translocation from the intracellular vesicles to the apical membrane of principal cells. Recently, serine-261 has been identified as a novel phosphorylation site on AQP2 and levels of phosphorylated S261 have been shown to decrease with vasopressin treatment suggesting its involvement in vasopressin-dependent AQP2 trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Aqp2
Gene Accession No. P34080, P41181, P56402, P79099
Gene Alias ADH water channel; AQP 2; AQP CD; Aqp2; AQP-2; AQPCD; AQP-CD; aquaporin 2; aquaporin 2 (collecting duct); Aquaporin CD; Aquaporin2; aquaporin-2; Aquaporin-CD; Aquaporine 2; collecting duct water channel protein; cph; jpk; MGC34501; Water channel protein for renal collecting duct; water-channel aquaporin 2; WCH CD; WCHCD; WCH-CD
Gene Symbols Aqp2
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminus portion of rat Aquaporin-2 (within residues 200-300).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100147810, 11827, 25386, 359, 539870
Target Species Human, Mouse, Rat, Bovine, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.