Learn More
Julabo™ Aqua Stabil™ Water Bath Protective Media
Product Code. 11930298
Change view
Click to view available options
:
Qty: 12 Bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11930298 Qty: 12 Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11930298 Supplier Julabo™ Supplier No. 8940012
Please to purchase this item. Need a web account? Register with us today!
Julabo™ Aqua Stabil™ Water Bath Protective Media stops growth of algae, bacteria and other microorganisms with its bactericidal effectiveness.
  • Protects bath fluid
  • Provides long period of use for fluid
Specifications
Description 12 Bottles
Type Aqua Stabil Protective Media
Product Identifier
  • Aqua-stabil
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.