missing translation for 'onlineSavingsMsg'
Learn More

APY0201, MedChemExpress

Product Code. 16439497
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439497 5 mg 5mg
16496948 10 mg 10mg
16487026 100 mg 100mg
16473007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16439497 Supplier Medchem Express Supplier No. HY15982

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress APY0201 is a potent PIKfyve inhibitor, which inhibits the conversion of PtdIns3P to PtdIns(3,5)P2 in the presence of in the presence of [33P]ATP with an IC50 of 5.2 nM. APY0201 also inhibits IL-12/IL-23 production.

APY0201 is a potent PIKfyve inhibitor, which inhibits the conversion of PtdIns3P to PtdIns(3,5)P2 in the presence of in the presence of [33P]ATP with an IC50 of 5.2 nM. APY0201 also inhibits IL-12/IL-23 production.

Specifications

Chemical Name or Material APY0201
Physical Form Solid
CAS 1232221-74-7
Color Earth Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H23N7O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 35 mg/mL (84.65 mM)
SMILES CC1=CC(/C=N/NC2=NC3=CC(C4=CC=NC=C4)=NN3C(N5CCOCC5)=C2)=CC=C1
Molecular Weight (g/mol) 413.48
Formula Weight 413.48
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.