missing translation for 'onlineSavingsMsg'
Learn More

APTO-253, MedChemExpress

Product Code. 16449567
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16449567

Brand: Medchem Express HY16291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress APTO-253 (LOR-253) is a small molecule that inhibits c-Myc expression, stabilizes G-quadruplex DNA, and induces cell cycle arrest and apoptosis in acute myeloid leukemia cells. APTO-253 mediates anticancer activity through induction of the Krüppel-like factor 4 (KLF4) tumor suppressor. APTO-253 has antiarthritic activity.

APTO-253 (LOR-253) is a small molecule that inhibits c-Myc expression, stabilizes G-quadruplex DNA, and induces cell cycle arrest and apoptosis in acute myeloid leukemia cells. APTO-253 mediates anticancer activity through induction of the Krüppel-like factor 4 (KLF4) tumor suppressor. APTO-253 has antiarthritic activity.

Specifications

Chemical Name or Material APTO-253
CAS 916151-99-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H14FN5
Quantity 5 mg
Synonym LOR-253 LT-253
Solubility Information DMSO : 60 mg/mL (163.32 mM; Need ultrasonic)
SMILES CC(N1)=C(C(N2)=NC3=C2C4=CC=CN=C4C5=NC=CC=C53)C6=C1C=CC(F)=C6
Molecular Weight (g/mol) 367.38
Formula Weight 367.38
Percent Purity 98.15%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.