Learn More
Novus Biologicals™ APRIL/TNFSF13 Antibody (BION-1301) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28382-50ug
 View more versions of this product
Product Code. 30046118
  • 213.00 € / 50µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
APRIL/TNFSF13 Monoclonal antibody specifically detects APRIL/TNFSF13 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
APRIL/TNFSF13
Monoclonal
Unconjugated
O75888
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG2
Flow Cytometry, ELISA, Functional Assay
BION-1301
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2
TNFSF13/ APRIL/ CD256
50 μg
Apoptosis, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology
8741
Human
Purified