Description
Includes
1 MicroAmp Centrifuge Adaptor
Accommodates a single MicroAmp reaction tube, fits a standard microcentrifuge rotor, and can be used at up to 15,000 x g.
- Accommodates single MicroAmp reaction tube for convenience
- Constructed to fit standard microcentrifuge rotor, offering flexibility
See user's manual or package insert for limited label license and trademark information.
PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store at Room Temperature |
| For Use With (Equipment) | 2720 Thermal Cycler, GeneAmp™ 9700, ProFlex™ PCR System, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti™ Thermal Cycler |
| Quantity | 1 Adaptor |