Learn More
Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer II (250 units/tube)
Shop All Applied Biosystems ProductsDescription
AmpliTaq DNA Polymerase is obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli., and is most thoroughly characterized enzyme available for PCR process. Its recombinant nature and purification method provide unparalleled purity and reproducibility.
- Characterized enzyme available for PCR process
- QC-tested to guarantee reproducible results
- Ideal for PCR
PCR, PCR and Real-Time PCR, Routine PCR
Specifications
Specifications
| Content And Storage | 1000 units of AmpliTaq DNA Polymerase and Buffer I. Store at –15 to –30°C. |
| Polymerase | AmpliTaq DNA Polymerase |
| Exonuclease Activity | 5' - 3' |
| Quantity | 1500 units |
| Fidelity (vs. Taq) | 1 X |
| Overhang | 3'-A |
| Size (Final Product) | 5 kb or less |