Learn More
Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer I (250 units/tube)
Product Code. 10486924 Shop All Applied Biosystems Products
Change view
Click to view available options
1500 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10486924 1500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10486924 Supplier Applied Biosystems™ Supplier No. N8080166
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A 94 kDa, thermostable, recombinant DNA polymerase
AmpliTaq™ DNA Polymerase is recombinant DNA polymerase obtained by expression of modified form of Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is thoroughly characterized enzyme available for PCR process and its recombinant nature and purification method provide unparalleled purity and reproducibility
  • Characterized enzyme available for the PCR process
  • QC-tested to guarantee reproducible results
  • Ideal for PCR
PCR, PCR and Real-Time PCR, Routine PCR
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 5 x 200μL of AmpliTaq DNA Polymerase (at 5U/μL); 4 x 1.5mL of GeneAmp 10X PCR Buffer (100mM Tris-HCl pH 8.3, 500mM KCl, 15mM MgCl2,0.01% (w/v) gelatin. Store at -20°C.
Polymerase AmpliTaq DNA Polymerase
Exonuclease Activity 5' - 3'
Quantity 1500 units
Fidelity (vs. Taq) 1 X
Overhang 3'-A
Size (Final Product) 5 kb or less