Learn More
APPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14294-1-AP-20UL
This item is not returnable.
View return policy
Description
Required for the regulation of cell proliferation in response to extracellular signals mediated by an early endosomal compartment. Links Rab5 to nuclear signal transduction.Specifications
| APPL2 | |
| Polyclonal | |
| Unconjugated | |
| APPL2 | |
| APPL2, Dip13 beta, DIP13B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 216190, 362860, 55198 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K3G9, Q8NEU8 | |
| Appl2 | |
| APPL2 Fusion Protein Ag5626 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |