Learn More
APPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14294-1-AP-20UL
 View more versions of this product
Product Code. 16812874
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Required for the regulation of cell proliferation in response to extracellular signals mediated by an early endosomal compartment. Links Rab5 to nuclear signal transduction.
Specifications
Specifications
APPL2
Polyclonal
Unconjugated
APPL2
APPL2, Dip13 beta, DIP13B
Rabbit
Antigen Affinity Chromatography
RUO
216190, 362860, 55198
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8K3G9, Q8NEU8
Appl2
APPL2 Fusion Protein Ag5626
20 μL
Primary
Mouse, Human, Rat