missing translation for 'onlineSavingsMsg'
Learn More

APPBP2 Antibody, Novus Biologicals™

Product Code. 18130162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18130162 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18130162 Supplier Novus Biologicals Supplier No. NBP215432

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

APPBP2 Polyclonal specifically detects APPBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APPBP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.44mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q92624
Gene Alias amyloid beta precursor protein (cytoplasmic tail) binding protein 2, Amyloid beta precursor protein-binding protein 2, amyloid protein-binding protein 2, APP-BP2, KIAA0228HS.84084, PAT1amyloid beta precursor protein (cytoplasmic tail)-binding protein 2, Protein interacting with APP tail 1
Gene Symbols APPBP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human APPBP2. The exact sequence is proprietary.
Molecular Weight of Antigen 67 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10513
Test Specificity Human. Expected to cross react based on sequence identity: Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.