Learn More
APP Antibody (6E10) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-62566-0.025MG
This item is not returnable.
View return policy
Description
APP Monoclonal specifically detects APP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy.
Specifications
| APP | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| APP | |
| Beta Amyloid signal peptide 1-16. | |
| 351 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunomicroscopy | |
| 60000000000 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Electron Microscopy 1:10 - 1:500 | |
| amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP | |
| Rabbit | |
| 0.025 mg | |
| This antibody binds to amino acid residues 1-16 of beta amyloid. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |