missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein D Antibody (APOD/3415) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14090JF585
This item is not returnable.
View return policy
Description
Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Specifications
| Apolipoprotein D | |
| Monoclonal | |
| Janelia Fluor 585 | |
| apo-D, ApoD, apolipoprotein D | |
| Human recombinant Apolipoprotein D protein fragment (around aa21-119) (exact sequence is proprietary) (Uniprot: P05090) | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 347 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| APOD/3415 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction