missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC3G Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77206AF700
This item is not returnable.
View return policy
Description
APOBEC3G Polyclonal antibody specifically detects APOBEC3G in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| APOBEC3G | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| APOBEC-related cytidine deaminase, APOBEC-related protein, APOBEC-related protein 9, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G, ARCD, ARP-9, bK150C2.7, CEM-15, CEM15ARP9, dJ494G10.1, DNA dC->dU-editing enzyme APOBEC-3G, EC 3.5.4, EC 3.5.4.-, EC 3.5.4.5, FLJ12740DNA dC->dU editing enzyme, MDS019, phorbolin-like protein MDS019 | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the amino-terminus of human APOBEC3G. The immunogen is located within the first 50 amino acids of APOBEC3G. Amino Acid Squence: RNTVERMYRDTFSYN | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 60489 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction