Learn More
Invitrogen™ APOBEC3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578800
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human CACO-2 whole cell, human A549 whole cell, rat PC-12 whole cell, mouse RAW264.7 whole cell .
PHO1 a member of the cytidine deaminase gene family. The PHO1 gene is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. This gene encodes a protein that lacks the zinc binding activity and may be an expressed pseudogene.
Specifications
| APOBEC3A | |
| Polyclonal | |
| Unconjugated | |
| APOBEC3A | |
| A3A; Apobec3; APOBEC3A; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3A; apolipoprotein B mRNA editing enzyme catalytic subunit 3A; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A; apolipoprotein B mRNA-editing complex 3; Arp3; BC003314; bK150C2.1; CEM15; CEM-15; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3A; pho1; PHO-1; phorbolin 1; Phorbolin1; phorbolin-1; PHRBN; probable DNA dC->dU-editing enzyme APOBEC-3A; recovery from Friend virus 3; Rfv3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200315, 80287 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 500 μg/mL | |
| Antibody with 0.9mg NaCl, 0.2mg Na2PO4, 4mg trehalose and 0.05mg sodium azide | |
| P31941, Q99J72 | |
| Apobec3, APOBEC3A | |
| E. coli-derived human PHO1 recombinant protein (Position: M1-L63). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.