missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
APLP2 May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5′-GTCACATG-3′ (CDEI box).
Specifications
Specifications
| Antigen | APLP2 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | APLP2 |
| Gene Accession No. | P15943, Q06335, Q06481 |
| Gene Alias | Amyloid like protein 2, Amyloid protein homolog, APLP 2, APLP2, APPH, APPL2, CDEBP, CDEI box binding protein |
| Gene Symbols | APLP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?