missing translation for 'onlineSavingsMsg'
Learn More

APLP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16864014 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16864014 20 μL 20µL
16854014 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16864014 Supplier Proteintech Supplier No. 150411AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

APLP2 May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5′-GTCACATG-3′ (CDEI box).

Specifications

Antigen APLP2
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene APLP2
Gene Accession No. P15943, Q06335, Q06481
Gene Alias Amyloid like protein 2, Amyloid protein homolog, APLP 2, APLP2, APPH, APPL2, CDEBP, CDEI box binding protein
Gene Symbols APLP2
Host Species Rabbit
Immunogen APLP2 Fusion Protein Ag6838
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11804, 334, 64312
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.