missing translation for 'onlineSavingsMsg'
Learn More

Apitolisib, MedChemExpress

Product Code. p-200096196
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496126

Brand: Medchem Express HY13246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Apitolisib (GDC-0980; GNE 390; RG 7422) is a selective, potent, orally bioavailable Class I PI3 kinase and mTOR kinase (TORC1/2) inhibitor with IC50s of 5 nM/27 nM/7 nM/14 nM for PI3Kα/PI3Kβ/PI3Kδ/PI3Kγ, and with a Ki of 17 nM for mTOR.

Apitolisib (GDC-0980

Specifications

Chemical Name or Material Apitolisib
Physical Form Solid
CAS 1032754-93-0
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C23H30N8O3S
For Use With (Application) Cancer-Kinase/protease
Synonym GDC-0980 GNE 390 RG 7422
Solubility Information DMSO : 14.29 mg/mL (28.66 mM; Need ultrasonic)
SMILES C[C@@H](C(N1CCN(CC1)CC2=C(C3=NC(C4=CN=C(N=C4)N)=NC(N5CCOCC5)=C3S2)C)=O)O
Molecular Weight (g/mol) 498.6
Formula Weight 498.6
Percent Purity 97.88%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.