missing translation for 'onlineSavingsMsg'
Learn More

Apigenin, MedChemExpress

Product Code. 16422396
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16422396

Brand: Medchem Express HYN1201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM.

Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM.

Specifications

Chemical Name or Material Apigenin
CAS 520-36-5
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C15H10O5
Quantity 500 mg
Synonym 4',5,7-Trihydroxyflavone Apigenol C.I. Natural Yellow 1
Solubility Information DMSO : 83.33 mg/mL (308.36 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=C2C(OC(C3=CC=C(O)C=C3)=CC2=O)=CC(O)=C1
Molecular Weight (g/mol) 270.24
Formula Weight 270.24
Percent Purity 96.92%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.