missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AP2M1 Polyclonal antibody specifically detects AP2M1 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | AP2M1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | Adaptin-mu2, adaptor-related protein complex 2, mu 1 subunit, AP-2 mu chain, Clathrin assembly protein complex 2 medium chain, clathrin coat adaptor protein AP50, Clathrin coat assembly protein AP50, Clathrin coat-associated protein AP50, clathrin-associated/assembly/adaptor protein, medium 1, HA2 50 kDa subunit, KIAA0109, mu subunit, Plasma membrane adaptor AP-2 50 kDa protein, plasma membrane adaptor AP-2 50kDA protein |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 134-433 of human AP2M1 (NP_001020376.1).,, Sequence:, TQQGIKSQTKEEQSQITSQVTGQIGWRREGIKYRRNELFLDVLESVNLLMSPQGQVLSAHVSGRVVMKSYLSGMPECKFGMNDKIVIEKQGKGTADETSKSGKQSIAIDDCTFHQCVRLSKFDSERSISFIPPDGEFELMRYRTTKDIILPFRVIPLVREVGRTKLEVKVVIKSNFKPSLLAQKIEVRIPTPLNTSGVQVICMKGKAKYKASENAIVWKIKRMAGMKESQISAEIELLPTNDKKKWARPPISMNFEVPFAPSGLKVRYLKVFEPKLNYSDHDVIKWVRYIGRSGIYETRC |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?