missing translation for 'onlineSavingsMsg'
Learn More

AP20187, MedChemExpress

Product Code. p-200098978
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498577

Brand: Medchem Express HY13992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AP20187 (B/B Homodimerizer) is a cell-permeable ligand used to dimerize FK506-binding protein (FKBP) fusion proteins and initiate biological signaling cascades and gene expression or disrupt protein-protein interactions.

AP20187 (B/B Homodimerizer) is a cell-permeable ligand used to dimerize FK506-binding protein (FKBP) fusion proteins and initiate biological signaling cascades and gene expression or disrupt protein-protein interactions.

Specifications

Chemical Name or Material AP20187
CAS 195514-80-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C82H107N5O20
Quantity 5 mg
Synonym B/B Homodimerizer
Solubility Information Ethanol : 150 mg/mL (101.16 mM; Need ultrasonic) ∣DMSO : ≥ 57 mg/mL (38.44 mM)
SMILES COC1=C(OC)C=C(CC[C@H](C2=CC=CC(OCC(NCC(CN(C)C)CNC(COC3=CC=CC([C@H](OC([C@H]4N(C([C@@H](CC)C5=CC(OC)=C(OC)C(OC)=C5)=O)CCCC4)=O)CCC6=CC(OC)=C(OC)C=C6)=C3)=O)=O)=C2)OC([C@@H]7CCCCN7C([C@H](C8=CC(OC)=C(OC)C(OC)=C8)CC)=O)=O)C=C1
Molecular Weight (g/mol) 1482.75
Formula Weight 1482.75
Percent Purity 95.33%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.