missing translation for 'onlineSavingsMsg'
Learn More

AP-III-a4 hydrochloride, MedChemExpress

Product Code. 16423506
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423506 25 mg 25mg
16462937 50 mg 50mg
16456878 10 mg 10mg
16449417 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16423506 Supplier Medchem Express Supplier No. HY15858A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AP-III-a4 (ENOblock) hydrochloride is a nonsubstrate analogue enolase inhibitor with an IC50 of 0.576 uM. AP-III-a4 hydrochloride can be used for the research of cancer and diabetic.

AP-III-a4 (ENOblock) hydrochloride is a nonsubstrate analogue enolase inhibitor with an IC50 of 0.576 uM. AP-III-a4 hydrochloride can be used for the research of cancer and diabetic.

Specifications

Chemical Name or Material AP-III-a4 hydrochloride
CAS 2070014-95-6
Color White
Purity Grade Notes Research
Molecular Formula C31H44ClFN8O3
Quantity 25 mg
Synonym ENOblock hydrochloride
Solubility Information DMSO : ≥ 53 mg/mL (83.97 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES FC1=CC=C(CNC2=NC(NCC3CCCCC3)=NC(NC4=CC=C(CC(NCCOCCOCCN)=O)C=C4)=N2)C=C1.Cl
Molecular Weight (g/mol) 631.18
Formula Weight 631.18
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.