Learn More
AP-III-a4 hydrochloride, MedChemExpress
Description
AP-III-a4 (ENOblock) hydrochloride is a nonsubstrate analogue enolase inhibitor with an IC50 of 0.576 uM. AP-III-a4 hydrochloride can be used for the research of cancer and diabetic.
Specifications
Specifications
| Chemical Name or Material | AP-III-a4 hydrochloride |
| CAS | 2070014-95-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H44ClFN8O3 |
| Quantity | 50 mg |
| Synonym | ENOblock hydrochloride |
| Solubility Information | DMSO : ≥ 53 mg/mL (83.97 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | FC1=CC=C(CNC2=NC(NCC3CCCCC3)=NC(NC4=CC=C(CC(NCCOCCOCCN)=O)C=C4)=N2)C=C1.Cl |
| Molecular Weight (g/mol) | 631.18 |
| Show More |