missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ANXA3 Recombinant Protein

335.00 € - 508.00 €

Specifications

Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Molecular Weight (g/mol) 61.27
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16101621
View Documents Promotion Details
Abnova™
H00000306-P01.10UG
10 ug
335.00 €
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16111621
View Documents Promotion Details
Abnova™
H00000306-P01.25UG
25μg
508.00 €
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
8
Glutathione Sepharose 4 Fast Flow
Human
Solution
61.27
In vitro wheat germ expression system
Wheat Germ (in vitro)
Recombinant
Videos
SDS
Documents

Documents

Product Certifications