Learn More
anti-Ubiquitin, Polyclonal, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34617JF549
Product Code. 18140164
  • 631.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
FGF-23 Monoclonal specifically detects FGF-23 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ubiquitin
Polyclonal
Q12841
Ubiquitin isolated from cow erythrocytes and conjugated to chicken gammaglobulins.
RUO
6233
Bovine, Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
0.5 mL
Primary
This antibody reacts with ubiquitin from human and several other mammalian species. It reacts with the periphery of senile plaques and neurofibrillary tangles in Alzheimer's disease. Cross-reacts with Human, Rat, Mouse and Cow. Not yet tested in other species.
Antisera