Learn More
anti-Ubiquitin, Polyclonal, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34617JF549
This item is not returnable.
View return policy
Description
FGF-23 Monoclonal specifically detects FGF-23 in Human samples. It is validated for ELISA.
Specifications
| Ubiquitin | |
| Polyclonal | |
| Q12841 | |
| Ubiquitin isolated from cow erythrocytes and conjugated to chicken gammaglobulins. | |
| RUO | |
| 6233 | |
| Bovine, Human, Mouse, Rat |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| 0.5 mL | |
| Primary | |
| This antibody reacts with ubiquitin from human and several other mammalian species. It reacts with the periphery of senile plaques and neurofibrillary tangles in Alzheimer's disease. Cross-reacts with Human, Rat, Mouse and Cow. Not yet tested in other species. | |
| Antisera |