Learn More
TP53RK, Human, Matched Antibody Pair (AP41), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00112858-AP41.1Set
Product Code. 16046979
  • 843.00 € / 1 set
Please to purchase this item. Need a web account? Register with us today!
Includes: Capture antibody: mouse monoclonal anti-TP53RK (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TP53RK, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
TP53RK
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TP53RK.
BUD32/C20orf64/Nori-2/Nori-2p/PRPK/dJ101A2
Mouse
RUO
112858
Human
Liquid
ELISA
Biotin
TP53RK
TP53RK
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TP53RK (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TP53RK, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications