missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP2A Mouse anti-Human, Clone: Ki-S1, Invitrogen™
Description
MA5-12433 targets Topoisomerase II alpha in IHC (P) applications and shows reactivity with Human samples. The MA5-12433 immunogen is lysate of the human histiocytic leukemia cell line U937.
Topo II alpha plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2- & M-phases in transformed and developmentally regulated normal cellsTopo II alpha is also implicated in drug resistance of tumor cells.
Specifications
Specifications
| Antigen | TOP2A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | Ki-S1 |
| Conjugate | Unconjugated |
| Formulation | tissue culture supernatant with 0.09% sodium azide |
| Gene | TOP2A |
| Gene Accession No. | P11388 |
| Gene Alias | DNA gyrase, DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, OTTHUMP00000207108 |
| Gene Symbols | TOP2A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?