missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-TNP, Alexa Fluor 488, Clone: G155-228, BD
Brand: BD Biosciences 562409
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: G155-228
Isotype: IgM, κ
Species Reactivity: Rat
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
| TNP | |
| G155-228 | |
| Alexa Fluor 488 | |
| TNP-keyhole limpet hemocyanin | |
| 0.1 mg | |
| Primary | |
| IgM κ |
| Monoclonal | |
| 0.2mg/mL | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction