Learn More
Ter-119 Magnetic Particles - DM Rat anti-Mouse, BD IMag-DM, Clone: TER-119, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 558536
Description
The TER-119 antibody reacts with a 52kDa molecule associated with glycophorin A on cells of the erythroid lineage in embryonic yolk sac, fetal liver, newborn liver, adult bone marrow, adult peripheral blood, and adult lymphoid organs. The TER-119 antigen is expressed on erythroid cells from pro-erythroblast through mature erythrocyte stages, but not on cells with BFU-E or CFU-E activities. The TER-119 epitope is not detected on hematopoietic stem cells, lymphoid cells, myeloid cells, or erythroleukemia lines. The TER-119 mAb is a component of the “lineage cocktail” used in studies of hematopoietic progenitors to detect, or deplete cells committed to the hematopoietic lineages.
Cell Separation
Specifications
| Ter-119 Magnetic Particles - DM | |
| Monoclonal | |
| BD IMag-DM | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| TER-119 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse Fetal Liver | |
| 5 mL | |
| Immunology | |
| Mouse | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.