missing translation for 'onlineSavingsMsg'
Learn More

transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A) (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16186335
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16186335 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16186335 Supplier Abnova Supplier No. H00006924B01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human TCEB3 protein.

This gene encodes the protein elongin A, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. [provided by RefSeq

Sequence: MAAESALQVVEKLQARLAANPDPKKLLKYLKKLSTLPITVDILAETGVGKTVNSLRKHEHVGSFARDLVAQWKKLVPVERNAEPDEQDFEKSNSRKRPRDALQKEEEMEGDYQETWKATGSRSYSPDHRQKKHRKLSELERPHKVSHGHERRDERKRCHRMSPTYSSDPESSDYGHVQSPPSCTSPHQMYVDHYRSLEEDQEPIVSHQKPGKGHSNAFQDRLGASQERHLGEPHGKGVVSQNKEHKSSHKDKRPVDAKSDEKASVVSREKSHKALSKEENRRPPSGDNAREKPPSSGVKKEKDREGSSLKKKCLPPSEAASDNHLKKPKHRDPEKAKLDKSKQGLDSFDTGKGAGDLLPKVKEKGSNNLKTPEGKVKTNLDRKSLGSLPKVEETDMEDEFEQPTMSFESYLSYDQPRKKKKKIVKTSATALGDKGLKKNDSKSTGKNLDSVQKLPKVNKTKSEKPAGADLAKLRKVPDVLPVLPDLPLPAIQANYRPLPSLELISSFQPKRKAFSSPQEEEEAGFTGRRMNSKMQVYSGSKCAYLPKMMTLHQQCIRVLKNNIDSIFEVGGVPYSVLEPVLERCTPDQLYRIEEYNHVLIEETDQLWKVHCHRDFKEERPEEYESWREMYLRLQDAREQRLRVLTKNIQFAHANKPKGRQAKMAFVNSVAKPPRDVRRRQEKFGTGGAAVPEKIKIKPAPYPMGSSHASASSISFNPSPEEPAYDGPSTSSAHLAPVVSSTVSYDPRKPTVKKIAPMMAKTIKAFKNRFSRR

Specifications

Antigen transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TCEB3 protein.
Formulation No additive
Gene TCEB3
Gene Accession No. NM_003198.1
Gene Alias FLJ38760/FLJ42849/SIII/TCEB3A
Gene Symbols TCEB3
Host Species Mouse
Immunogen TCEB3 (NP_003189.1, 1 a.a. ∼ 772 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6924
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.