missing translation for 'onlineSavingsMsg'
Learn More

TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16115696
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16115696 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16115696 Supplier Abnova Supplier No. H00027097D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against a full-length human TAF5L protein.

The product of this gene belongs to the WD-repeat TAF5 family of proteins. This gene encodes a protein that is a component of the PCAF histone acetylase complex. The PCAF histone acetylase complex, which is composed of more than 20 polypeptides some of which are TAFs, is required for myogenic transcription and differentiation. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors to facilitate complex assembly and transcription initiation. The encoded protein is structurally similar to one of the histone-like TAFs, TAF5. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq

Sequence: MKRVRTEQIQMAVSCYLKRRQYVDSDGPLKQGLRLSQTAEEMAANLTVQSESGCANIVSAAPCQAEPQQYEVQFGRLRNFLTDSDSQHSHEVMPLLYPLFVYLHLNLVQNSPKSTVESFYSRFHGMFLQNASQKDVIEQLQTTQTIQDILSNFKLRAFLDNKYVVRLQEDSYNYLIRYLQSDNNTALCKVLTLHIHLDVQPAKRTDYQLYASGSSSRSENNGLEPPDMPSPILQNEAALEVLQESIKRVKDGPPSLTTICFYAFYNTEQLLNTAEISPDSKLLAAGFDNSCIKLWSLRSKKLKSEPHQVDVSRIHLACDILEEEV

Specifications

Antigen TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TAF5L protein.
Formulation PBS with no preservative; pH 7.4
Gene TAF5L
Gene Accession No. NM_001025247.1
Gene Alias PAF65B
Gene Symbols TAF5L
Host Species Rabbit
Immunogen TAF5L (NP_001020418.1, 1 a.a. ∼ 325 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 27097
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.