missing translation for 'onlineSavingsMsg'
Learn More

GPR91, Polyclonal, Invitrogen™

Product Code. 13227898
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13227898 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13227898 Supplier Invitrogen Supplier No. PA523299

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SUCNR1 is a 330 amino acid containing membrane receptor, belonging to the G-protein coupled receptor 1 family, characterized by an extracellular N terminus, seven transmembrane regions, and an intracellular C terminus and playing a role in cell communication. SUCNR1 a receptor for the citric acid cycle intermediate succinate, functions in establishing a neovascular network during development and in response to injury. SUCNR1, in response to increased succinate levels, regulates the production of numerous angiogenic factors including VEGF and may be involved in the modulation of platelet function. It is known to play an essential role in proliferative ischemic retinopathy and is an attractive target for treating diseases such as diabetic retinopathy that stem from aberrant neovascularization in the mature retina. Expression of SUCNR1 is common in kidney and in some other tissues.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SUCNR1/GPR91
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Gene SUCNR1
Gene Accession No. Q9BXA5
Gene Alias GPR91, G protein-coupled receptor 91, G-protein coupled receptor 91, P2Y purinoceptor 1
Gene Symbols SUCNR1
Host Species Rabbit
Immunogen Synthetic peptide (121 KYPFREHLLQKKE 133) of SUCNR1/GPR91 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 56670
Target Species Human, Non-human Primate
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.