Learn More
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SPOCK3.
Brand: Abnova H00050859-A01.50uL
Description
Proteoglycans, which consist of a core protein and covalently linked glycosaminoglycans, are components of the extracellular matrix. SPOCK3 encodes a member of a novel Ca(2+)-binding proteoglycan family.[supplied by OMIM
Sequence: RGPILSTCKQCPVVYPSPVCGSDGHTYSFQCKLEYQACVLGKQISVKCEGHCPCPSDKPTSTSRNVKRACSDLEFREVANRLRDWFKALHESGSQNKKTSpecifications
| sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SPOCK3. | |
| SPOCK3 | |
| HSAJ1454/TES-3 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 50859 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_016950 | |
| SPOCK3 | |
| SPOCK3 (NP_058646, 132 a.a. ∼ 230 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.