Learn More
surfactant protein C, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SFTPC protein.
Brand: Abnova H00006440-D01P.100ug
Description
The SFTPC gene encodes pulmonary-associated surfactant protein C (SPC), an extremely hydrophobic surfactant protein essential for lung function and homeostasis after birth. It is produced exclusively by type II alveolar epithelial cells in the lung. Pulmonary surfactant is a lipid-rich material that prevents lung collapse by lowering surface tension at the air-liquid interface in the alveoli of lung. Surfactant is composed of phospholipids and other surfactant-associated proteins (Clark and Clark, 2005 [PubMed 15927881]). See also SFTPA1 (MIM 178630), SFTPB (MIM 178640), and SFTPD (MIM 178635).[supplied by OMIM
Sequence: MDVGSKEVLMESPPDYSAAPRGRFGIPCCPVHLKRLLIVVVVVVLIVVVIVGALLMGLHMSQKHTEMVLEMSIGAPEAQQRLALSEHLVTTATFSIGSTGLVVYDYQQLLIAYKPAPGTCCYIMKIAPESIPSLEALNRKVHNFQMECSLQAKPAVPTSKLGQAEGRDAGSAPSGGDPAFLGMAVNTLCGEVPLYYISpecifications
| surfactant protein C | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human SFTPC protein. | |
| SFTPC | |
| PSP-C/SFTP2/SMDP2/SP-C | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6440 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003018 | |
| SFTPC | |
| SFTPC (NP_003009.1, 1 a.a. ∼ 197 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.