missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNase12 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15574381
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15574381 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574381 Supplier Invitrogen™ Supplier No. PA537411

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 17 kDa. Purity is >95% by SDS-PAGE.

Probable inactive ribonuclease-like protein 12, RNASE 12, belongs to the pancreatic ribonuclease family and does not exhibit any ribonuclease activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNase12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene RNASE12
Gene Accession No. Q5GAN4
Gene Alias epididymis secretory sperm binding protein Li 85p; HEL-S-85p; Probable inactive ribonuclease-like protein 12; Rai1; RGD1307829; ribonuclease 12; ribonuclease A family member 12 (inactive); ribonuclease A I1; ribonuclease, RNase A family, 12 (non-active); ribonuclease-like protein 12; RNASE12; RNS12
Gene Symbols RNASE12
Host Species Rabbit
Immunogen Recombinant full length human RNASE12.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493901
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.