Learn More
reversion-inducing-cysteine-rich protein with kazal motifs, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RECK protein.
Brand: Abnova H00008434-B01.50uL
Description
The protein encoded by this gene is a cysteine-rich, extracellular protein with protease inhibitor-like domains whose expression is suppressed strongly in many tumors and cells transformed by various kinds of oncogenes. In normal cells, this membrane-anchored glycoprotein may serve as a negative regulator for matrix metalloproteinase-9, a key enzyme involved in tumor invasion and metastasis. [provided by RefSeq
Sequence: MATVRASLRGALLLLLAVAGVAEVAGGLAPGSAGALCCNHSKDNQMCRDVCEQIFSSKSESRLKHLLQRAPDYCPETMVEIWNCMNSSLPGVFKKSDGWVGLGCCELAIALECRQACKQASSKNDISKVCRKEYENALFSCISRNEMGSVCCSYAGHHTNCREYCQAIFRTDSSPGPSQIKAVENYCASISPQLIHCVNNYTQSYPMRNPTDSLYCCDRAEDHACQNACKRILMSKKTEMEIVDGLIEGCKTQPLPQDPLWQCFLESSQSVHPGVTVHPPPSTGLDGAKLHCCSKANTSTCRELCTKLYSMSWGNTQSWQEFDRFCEYNPVEVSMLTCLADVREPCQLGCRNLTYCTNFNNRPTELFRSCNAQSDQGAMNDMKLWEKGSIKMPFINIPVLDIKKCQPEMWKAIACSLQIKPCHSKSRGSIICKSDCVEILKKCGDQNKFPEDHTAESICELLSPTDDLKNCIPLDTYLRPSTLGNIVEEVTHPCNPNPCPANELCEVNRKGCPSGDPCLPYFCVQGCKLGEASDFIVRQGTLIQVPSSAGEVGCYKICSCGQSGLLENCMEMHCIDLQKSCIVGGKRKSHGTSFSIDCNVCSCFAGNLVCSTRLCLSEHSSEDDRRTFTGLPCNCADQFVPVCGQNGRTYPSACIARCVGLQDHQFEFGSCMSKDPCNPNPCQKNQRCIPKPQVCLTTFDKFGCSQYECVPRQLACDQVQDPVCDTDHMEHNNLCTLYQRGKSLSYKGPCQPFCRATEPVCGHNGETYSSVCAAYSDRVAVDYYGDCQAVGVLSEHSSVAECASVKCPSLLAAGCKPIIPPGACCPLCAGMLRVLFDKEKLDTIAKVTNKKPITVLEILQKIRMHVSVPQCDVFGYFSIESEIVILIIPVDHYPKALQIEACNKEAEKIESLINSDSPTLASHVPLSALIISQVQVSSSVPSAGVRARPSCHSLLLPLSLGLALHLLWTYNSpecifications
| reversion-inducing-cysteine-rich protein with kazal motifs | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human RECK protein. | |
| RECK | |
| ST15/hRECK | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_021111 | |
| RECK | |
| RECK (NP_066934, 1 a.a. ∼ 971 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 8434 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.