Learn More
Phosphoserine/threonine Mouse, Unlabeled, Clone: 22A, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 612549
Description
Protein phosphorylation of serine and threonine residues is critical for the control of protein activity involved in various cellular events. An assortment of Ser/Thr kinases and phosphatases regulate serine and threonine phosphorylation in cell signaling pathways, such as growth factor, cytokine, chemokine, and stress response. Detection of serine and threonine phosphorylation can generally be monitored by antibodies that detect phosphoserine and phosphothreonine.
Immunofluorescence, Western Blotting
Specifications
| Phosphoserine/threonine | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 22A | |
| Unconjugated | |
| Mouse | |
| 150 μg | |
| Cell Biology | |
| Human, Rat | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.