Learn More
nuclear receptor subfamily 1, group D, member 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human NR1D2 protein.
Brand: Abnova H00009975-D01P.100ug
Description
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described. [provided by RefSeq
Sequence: MEVNAGGVIAYISSSSSASSHASCHSEGSENSFQSSSSSVPSSPNSSNSDTNGNPKNGDLANIEGILKNDRIDCSMKTSKSSAPGMTKSHSGVTKFSGMVLLCKVCGDVASGFHYGVHACEGCKGFFRRSIQQNIQYKKCLKNENCSIMRMNRNRCQQCRFKKCLSVGMSRDAVRFGRIPKREKQRMLIEMQSAMKTMMNSQFSGHLQNDTLVEHHEQTALPAQEQLRPKPQLEQENIKSSSPPSSDFAKEEVIGMVTRAHKDTFMYNQEQQENSAESMQPKRGERIRKNMEQYNLNHDHCGNGLSSHFPCSESQQHLNGQFKGRNIMHYPNGHAICIANGHCMNFSNAYTQRVCDRVPIDGFSQNENKNSYLCNTGGRMHLVCPMSKSPYVDPHKSGHEIWEEFSMSFTPAVKEVVEFAKRIPGFRDLSQHDQVNLLKAGTFEVLMVRFASLFDAKERTVTFLSGKKYSVDDLHSMGAGDLLNSMFEFSEKLNALQLSDEEMSLFTAVVLVSADRSGIENVNSVEALQETLIRALRTLIMKNHPNEASIFTKLLLKLPDLRSLNNMHSEELLAFKVHPSpecifications
| nuclear receptor subfamily 1, group D, member 2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human NR1D2 protein. | |
| NR1D2 | |
| BD73/EAR-1r/HZF2/Hs.37288/RVR | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005126.2 | |
| NR1D2 | |
| NR1D2 (NP_005117.2, 1 a.a. ∼ 579 a.a) full-length human protein. | |
| 100 μg | |
| Transcription Regulation | |
| 9975 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.