missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-NQO1 Matched Antibody Pair, Abnova™
Brand: Abnova H00001728-AP11.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-NQO1 (100μg; Detection antibody: mouse monoclonal anti-NQO1, IgG1 Kappa (20μg; Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Host Species: Rabbit, Mouse
Isotype: IgG, IgGk1, κ
Cross Reactivity: Human
ELISA Pair
Specifications
NQO1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NQO1. | |
DHQU/DIA4/DTD/NMOR1/NMORI/QR1 | |
Mouse, Rabbit | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-NQO1 (100 μg) 2. Detection antibody: mouse monoclonal anti-NQO1, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
NQO1 | |
NQO1 | |
1 Set | |
1728 | |
Human | |
IgG |