Learn More
NLRP3/NALP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12446
Description
NLRP3/NALP3 Polyclonal specifically detects NLRP3/NALP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunomicroscopy, Knockdown Validated.Specifications
| NLRP3/NALP3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, anti-mouse NLRP3, anti-NLRP3, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, NLRP3 mouse, NLRP3 polyclonal, NLRP3 rat, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 | |
| Rabbit | |
| 118 kDa | |
| 0.2 mL | |
| Primary | |
| Reacts with Human, Mouse, New World Monkey and Rat. The amino acid sequence used as immunogen is 100% homologous in Human and Monkey and 86% homologous in Mouse and Rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown, KnockDown | |
| 1.0 mg/mL | |
| Western Blot 2 to 5 μg/mL, Immunohistochemistry 1:10 to 1:50, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10 to 1:50, Immunomicroscopy, Knockdown Validated | |
| Q96P20 | |
| NLRP3 | |
| This NLRP3/NALP3 antibody was raised against a portion of amino acids 1-50 of human NLRP3/NALP3. | |
| Immunogen affinity purified | |
| RUO | |
| 114548 | |
| Human, Mouse, Rat | |
| Purified |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only