Learn More
NIMA (never in mitosis gene a)- related kinase 8, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human NEK8 protein.
Brand: Abnova H00284086-B01P.50ug
Description
This gene encodes a member of the serine/threionine protein kinase family related to NIMA (never in mitosis, gene A) of Aspergillus nidulans. The encoded protein may play a role in cell cycle progression from G2 to M phase. Mutations in the related mouse gene are associated with a disease phenotype that closely parallels the juvenile autosomal recessive form of polycystic kidney disease in humans. [provided by RefSeq
Sequence: MEKYERIRVVGRGAFGIVHLCLRKADQKLVIIKQIPVEQMTKEERQAAQNECQVLKLLNHPNVIEYYENFLEDKALMIAMEYAPGGTLAEFIQKRCNSLLEEETILHFFVQILLALHHVHTHLILHRDLKTQNILLDKHRMVVKIGDFGISKILSSKSKAYTVVGTPCYISPELCEGKPYNQKSDIWALGCVLYELASLKRAFEAANLPALVLKIMSGTFAPISDRYSPELRQLVLSLLSLEPAQRPPLSHIMAQPLCIRALLNLHTDVGSVRMRRAEKSVAPSNTGSRTTSVRCRGIPRGPVRPAIPPPLSSVYAWGGGLGTPLRLPMLNTEVVQVAAGRTQKAGVTRSGRLILWEAPPLGAGGGSLLPGAVEQPQPQFISRFLEGQSGVTIKHVACGDFFTACLTDRGIIMTFGSGSNGCLGHGSLTDISQPTIVEALLGYEMVQVACGASHVLALSTERELFAWGRGDSGRLGLGTRESHSCPQQVPMPPGQEAQRVVCGIDSSMILTVPGQALACGSNRFNKLGLDHLSLGEEPVPHQQVEEALSFTLLGSAPLDQEPLLSIDLGTAHSAAVTASGDCYTFGSNQHGQLGTNTRRGSRAPCKVQGLEGIKMAMVACGDAFTVAIGAESEVYSWGKGARGRLGRRDEDAGLPRPVQLDETHPYTVTSVSCCHGNTLLAVRSVTDEPVPPSpecifications
| NIMA (never in mitosis gene a)- related kinase 8 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human NEK8 protein. | |
| NEK8 | |
| JCK/MGC138445/NEK12A | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_178170.2 | |
| NEK8 | |
| NEK8 (NP_835464.1, 1 a.a. ∼ 692 a.a) full-length human protein. | |
| 50 μg | |
| Kinases | |
| 284086 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.