missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOD Monoclonal Antibody (5.2F), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512907
This item is not returnable.
View return policy
Description
MA5-12907 targets MyoD1 in IF, IM, and IP applications and shows reactivity with Chicken, Human, mouse, and Rat samples. The MA5-12907 immunogen is recombinant mouse MyoD1 protein.
The Myogenic determination gene (MyoD) was first identified by the virtue of its ability to convert embryonic mouse fibroblast cells to muscle cells. It was subsequently shown that forced expression of MyoD (human homolog is myf 3) gene in a wide variety of normal and neoplastic cells could either convert the cells to muscle cells or activate a set of the otherwise transcriptionally inactive muscle-specific genes in these cells. The regulatory domain of the MyoD gene product lies within a 70 amino acid region and comprises a basic DNA binding motif and a helix-loop-helix (HLH) dimerization motif. Subsequent studies identified three other genes whose products shared sequence homology for the basic HLH domain of MyoD. These are; myf5, myogenin (human homolog is myf4) and myf6 (also known as MRF4 and herculin).
Specifications
| MYOD | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P10085, P15172, P16075, Q02346 | |
| MYOD1 | |
| Recombinant mouse MyoD1 protein | |
| 500 μL | |
| Primary | |
| Chicken, Human, Mouse, Rat | |
| Antibody | |
| IgG2a κ |
| Immunocytochemistry, Immunomicroscopy, Immunoprecipitation | |
| 5.2F | |
| Unconjugated | |
| MYOD1 | |
| AI503393; bHLHc1; Class C basic helix-loop-helix protein 1; CMD1; MYF3; Myf-3; Myoblast determination protein 1; myoblast determination protein 1 homolog; Myod; MYOD1; Myod-1; MYOD1 homolog; myogenic differentiation 1; myogenic factor 1; myogenic factor 3; PUM; unnamed protein product | |
| Mouse | |
| Protein A | |
| RUO | |
| 17927, 337868, 374048, 4654 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction