missing translation for 'onlineSavingsMsg'
Learn More

anti-Mouse IgM, κ Isotype Control, Clone: G155-228, BD

Product Code. 15802169
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15802169 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15802169 Supplier BD Biosciences Supplier No. 562704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The G155-228 antibody is specific for the hapten trinitrophenol (TNP), a hapten not expressed on human, non-human primate, mouse, or rat primary cells or cell lines. In the absence of specific binding, this immunoglobulin may bind non-specifically to Fc receptors.

Host Species: Mouse
Clone: G155-228
Isotype: IgM, κ

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone G155-228
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen TNP-Keyhole Limpet Hemocyanin
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.