missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516287
This item is not returnable.
View return policy
Description
This antibody labels basement membrane laminin proteins surrounding normal vessels, glands, muscles, nerves, fat and decidual cells separating these from abutting connective tissues.
Extracellular matrix (ECM) surrounds and separates most tissue compartments, determines tissue architecture and dynamically interacts with its surrounding cells. The basement membrane is a part of the ECM and is composed of collagens, proteoglycans, elastin and several glycoproteins of which laminin is the most abundant. Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains Dystroglycan is a major laminin receptor in muscle and epithelia, and nidogen is a high-affinity laminin-binding protein important for basement-membrane assembly.
Specifications
| Laminin | |
| Polyclonal | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P25391 | |
| LAMA1 | |
| Laminin isolated from a rat yolk sac tumor cell line | |
| 500 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| LAMA1 | |
| S-LAM alpha, S-laminin subunit alpha, laminin A chain, laminin subunit alpha-1, laminin-3 subunit alpha | |
| Rabbit | |
| Purified | |
| RUO | |
| 284217, 316758 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction