Learn More
KIR2DS4 (CD158i) Mouse anti-Human, Alexa Fluor 647, Clone: 179315, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 564375
Description
The 179315 monoclonal antibody specifically binds to Killer cell immunoglobulin-like receptor 2DS4 which is also known as CD158i, KKA3, and NKAT8. CD158i is encoded by KIR2DS4 (killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4). It is a type I transmembrane glycoprotein that can bind to HLA-C antigens and function as an activating immunoglobulin-like receptor that is expressed on NK cells and some T cells.
Specifications
| KIR2DS4 (CD158i) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| 179315 | |
| KIR2DS4; KIR antigen 2DS4; KKA3; NKAT8; p58 NK receptor CL-39/CL-1 | |
| Human KIR2DS4-Transfected Cell Line | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.